siRNA Details
| Gene Symbol: Snrpa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021336.4 | NP_067311.4 | 5 | 652 | ||
| GenBank mRNA | AF230356.1 | - | 5 | 381 | ||
| AK005256.1 | - | 5 | 444 | |||
| AK088438.1 | - | 5 | 427 | |||
| AK151800.1 | - | 5 | 445 | |||
| AK152546.1 | - | 5 | 445 | |||
| AK153502.1 | - | 5 | 445 | |||
| AK206837.1 | - | 4 | 215 | |||
| BC013777.1 | - | 5 | 419 | 2651704 | ||
| BC029642.1 | - | 5 | 419 | 3588138 | ||

Snrpa1
Genomic Map