siRNA Details
| Gene Symbol: Nde1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001285503.1 | NP_001272432.1 | 9 | 1679 | ||
| NM_001285504.1 | NP_001272433.1 | 8 | 1402 | |||
| NM_023317.2 | NP_075806.2 | 9 | 1551 | |||
| GenBank mRNA | AF290473.1 | - | 7 | 1286 | ||
| AF322073.1 | - | 8 | 1458 | |||
| AK012830.1 | - | 9 | 1481 | |||
| AK012861.1 | - | 9 | 1499 | |||
| AK037289.1 | - | 9 | 1494 | |||
| AK146982.1 | - | 9 | 1609 | |||
| AK166858.1 | - | 8 | 1381 | |||
| AK169743.1 | - | 9 | 1481 | |||
| BC023267.1 | - | 9 | 1530 | 3672565 | ||

Nde1
Genomic Map