siRNA Details
| Gene Symbol: 2210010C04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023333.4 | NP_075822.3 | 5 | 1170 | ||
| GenBank mRNA | AK008695.1 | - | 5 | 1169 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2210010C04Rik| Gene Symbol: 2210010C04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023333.4 | NP_075822.3 | 5 | 1170 | ||
| GenBank mRNA | AK008695.1 | - | 5 | 1169 | ||