siRNA Details
| Gene Symbol: Adck3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163290.1 | NP_001156762.1 | 15 | 2212 | ||
| NM_023341.3 | NP_075830.2 | 15 | 2310 | |||
| GenBank mRNA | AJ278735.1 | - | 15 | 2190 | ||
| AK004177.1 | - | 8 | 1193 | |||
| AK004791.1 | - | 15 | 2252 | |||
| AK014605.2 | - | 15 | 2252 | |||
| AK192180.1 | - | 1 | 63 | |||
| AK202922.1 | - | 1 | 149 | |||
| AK219700.1 | - | 1 | 149 | |||
| BC030937.1 | - | 15 | 2258 | 4922892 | ||

Adck3
Genomic Map