siRNA Details
| Gene Symbol: SLU7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006425.4 | NP_006416.3 | 2 | 256 | ||
| XM_005265793.3 | XP_005265850.1 | 2 | 603 | |||
| XM_011534418.2 | XP_011532720.1 | 3 | 306 | |||
| GenBank mRNA | AF101074.1 | - | 1 | 101 | ||
| AK024875.1 | - | 2 | 176 | |||
| AK314564.1 | - | 2 | 176 | |||
| AY486334.1 | - | 2 | 136 | |||
| BC010634.1 | - | 2 | 182 | 3871882 | ||
| DA232975.1 | - | 2 | 258 | |||
| DA409854.1 | - | 2 | 182 | |||
| DQ174516.1 | - | 2 | 188 | |||
| DQ174517.1 | - | 2 | 226 | |||

SLU7
Genomic Map