siRNA Details
| Gene Symbol: Zfp788 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023363.2 | NP_075852.1 | 6 | 2705 | ||
| GenBank mRNA | AK013166.1 | - | 1 | 1803 | ||
| AK036346.1 | - | 4 | 2535 | |||
| AK082981.1 | - | 6 | 2706 | |||
| AK163311.1 | - | 6 | 2654 | |||
| BC027798.1 | - | 3 | 2434 | 5323247 | ||

Zfp788
Genomic Map