siRNA Details
| Gene Symbol: Rtp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023386.5 | NP_075875.3 | 2 | 1046 | ||
| GenBank mRNA | AJ251364.1 | - | 2 | 1041 | ||
| AK007477.1 | - | 2 | 1024 | |||
| AK018021.1 | - | 2 | 1039 | |||
| BC006590.1 | - | 1 | 624 | |||
| BC024872.1 | - | 2 | 1004 | 3990099 | ||

Rtp4
Genomic Map