siRNA Details
| Gene Symbol: Wfdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023395.2 | NP_075884.1 | 5, 6 | 764 | ||
| GenBank mRNA | AK132541.1 | - | 5, 6 | 765 | ||
| AK142707.1 | - | 5, 6 | 633 | |||
| AK161606.1 | - | 5, 6 | 630 | |||
| BC025850.1 | - | 5, 6 | 577 | |||
| BC038122.1 | - | 5, 6 | 606 | |||
| BC132073.1 | - | 5, 6 | 693 | 40130350 | ||
| BC132469.1 | - | 5, 6 | 693 | 40130746 | ||
| BC145305.1 | - | 5, 6 | 690 | 9053859 | ||

Wfdc1
Genomic Map