siRNA Details
| Gene Symbol: Mesdc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023403.3 | NP_075892.3 | 1 | 419 | ||
| GenBank mRNA | AK008735.1 | - | 1 | 136 | ||
| AK031949.1 | - | 1 | 162 | |||
| AK076760.1 | - | 1 | 159 | |||
| AK076773.1 | - | 1 | 154 | |||
| AK172009.1 | - | 1 | 159 | |||
| BC014742.1 | - | 1 | 140 | 4239248 | ||
| BC058612.1 | - | 3073 | ||||
| BU054434.1 | - | 1 | 419 | |||

Mesdc2
Genomic Map