siRNA Details
| Gene Symbol: Mum1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023431.5 | NP_075920.4 | 5 | 757 | ||
| GenBank mRNA | AK078792.1 | - | 2 | 464 | ||
| AK154610.1 | - | 5 | 676 | |||
| AK158750.1 | - | 5 | 757 | |||
| AK159972.1 | - | 4 | 1659 | |||
| AK170459.1 | - | 5 | 676 | |||
| AK210750.1 | - | 1 | 322 | |||
| BC008550.1 | - | 2 | 433 | |||
| BC075617.1 | - | 4 | 646 | 5718202 | ||

Mum1
Genomic Map