siRNA Details
Gene Symbol: Tmem132e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC137906.1 | - | 2, 3 | 1061 | 8860925 | |
BC145026.1 | - | 2, 3 | 1061 | 9053574 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tmem132e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC137906.1 | - | 2, 3 | 1061 | 8860925 | |
BC145026.1 | - | 2, 3 | 1061 | 9053574 |