siRNA Details
| Gene Symbol: 1110032A03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023483.3 | NP_075972.2 | 4 | 1482 | ||
| GenBank mRNA | AK043513.1 | - | 4 | 1483 | ||
| AK158460.1 | - | 5 | 1446 | |||
| AK161835.1 | - | 90 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1110032A03Rik| Gene Symbol: 1110032A03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023483.3 | NP_075972.2 | 4 | 1482 | ||
| GenBank mRNA | AK043513.1 | - | 4 | 1483 | ||
| AK158460.1 | - | 5 | 1446 | |||
| AK161835.1 | - | 90 | ||||