siRNA Details
| Gene Symbol: Mrps9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023514.4 | NP_076003.3 | 9 | 931 | ||
| GenBank mRNA | AJ250193.1 | - | 5 | 400 | ||
| AK009082.1 | - | 9 | 881 | |||
| AK049465.1 | - | 8 | 1920 | |||
| AK144656.1 | - | 9 | 889 | |||
| AK150985.1 | - | 9 | 888 | |||
| BC022587.1 | - | 4 | 369 | |||
| BC051936.1 | - | 8 | 624 | |||
| BC069980.1 | - | 7 | 525 | |||
| BC116437.2 | - | 9 | 881 | 40104493 | ||

Mrps9
Genomic Map