siRNA Details
| Gene Symbol: Rab3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023537.5 | NP_076026.1 | 4 | 544 | ||
| GenBank mRNA | AB232588.1 | - | 3 | 395 | ||
| AF312036.1 | - | 3 | 395 | |||
| AK012165.1 | - | 4 | 504 | |||
| AK082959.1 | - | 4 | 544 | |||
| BC051918.1 | - | 4 | 449 | 5691723 | ||
| BC057173.1 | - | 3 | 413 | 4989309 | ||

Rab3b
Genomic Map