siRNA Details
| Gene Symbol: Rab3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023537.5 | NP_076026.1 | 5 | 1947 | ||
| GenBank mRNA | AK082959.1 | - | 5 | 1947 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rab3b| Gene Symbol: Rab3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023537.5 | NP_076026.1 | 5 | 1947 | ||
| GenBank mRNA | AK082959.1 | - | 5 | 1947 | ||