siRNA Details
| Gene Symbol: Jam2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023844.5 | NP_076333.3 | 3, 4 | 616 | ||
| GenBank mRNA | AF255911.1 | - | 3, 4 | 339 | ||
| AJ291757.1 | - | 3, 4 | 552 | |||
| AK010616.1 | - | 3, 4 | 458 | |||
| AK010826.1 | - | 3, 4 | 460 | |||
| AK013914.1 | - | 3, 4 | 457 | |||
| AK028757.1 | - | 3, 4 | 307 | |||
| AK045095.1 | - | 3, 4 | 354 | |||
| AK153636.1 | - | 3, 4 | 306 | |||
| BC028778.1 | - | 3, 4 | 616 | 1195543 | ||

Jam2
Genomic Map