siRNA Details
| Gene Symbol: Rab3c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023852.5 | NP_076341.1 | 5 | 929 | ||
| GenBank mRNA | AK014132.1 | - | 5 | 851 | ||
| AK079574.1 | - | 5 | 826 | |||
| AK148904.1 | - | 5 | 850 | |||
| AK161006.1 | - | 5 | 827 | |||
| AK164316.1 | - | 5 | 874 | |||
| AY026947.1 | - | 5 | 810 | |||
| BC114335.2 | - | 5 | 802 | 40045744 | ||

Rab3c
Genomic Map