siRNA Details
| Gene Symbol: Ndufa10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024197.1 | NP_077159.1 | 2 | 199 | ||
| GenBank mRNA | AK005339.1 | - | 2 | 235 | ||
| AK013675.1 | - | 2 | 243 | |||
| AK046292.1 | - | 103 | ||||
| AK145979.1 | - | 2 | 255 | |||
| AK177419.1 | - | 1 | 53 | |||
| AK184477.1 | - | 1 | 53 | |||
| AK185477.1 | - | 1 | 53 | |||
| BC003439.1 | - | 2 | 199 | 2647815 | ||

Ndufa10
Genomic Map