siRNA Details
| Gene Symbol: Fam120b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024203.3 | NP_077165.2 | 3 | 1829 | ||
| GenBank mRNA | AK016568.1 | - | 3 | 1795 | ||
| AK044023.1 | - | 1 | 2480 | |||
| AK132211.1 | - | 3 | 1807 | |||
| AK220425.1 | - | 2 | 4416 | |||
| BC003200.1 | - | 2 | 732 | |||
| BC066088.1 | - | 3 | 1874 | |||
| BC085163.1 | - | 3 | 3095 | 30618952 | ||
| DQ873694.1 | - | 3 | 1810 | |||

Fam120b
Genomic Map