siRNA Details
| Gene Symbol: Zfp593 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024215.2 | NP_077177.2 | 3 | 1929 | ||
| GenBank mRNA | AK133585.1 | - | 3 | 1926 | ||
| AK156931.1 | - | 3 | 1867 | |||
| BC003465.1 | - | 3 | 1870 | 2648747 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp593| Gene Symbol: Zfp593 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024215.2 | NP_077177.2 | 3 | 1929 | ||
| GenBank mRNA | AK133585.1 | - | 3 | 1926 | ||
| AK156931.1 | - | 3 | 1867 | |||
| BC003465.1 | - | 3 | 1870 | 2648747 | ||