siRNA Details
| Gene Symbol: Cmtm3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024217.3 | NP_077179.1 | 5 | 1260 | ||
| GenBank mRNA | AK003882.1 | - | 5 | 1116 | ||
| AK079165.1 | - | 5 | 1229 | |||
| AK172091.1 | - | 5 | 1163 | |||
| AK198392.1 | - | 1 | 355 | |||
| AY241870.1 | - | 5 | 1254 | |||
| BC003230.1 | - | 5 | 1167 | 3483351 | ||

Cmtm3
Genomic Map