siRNA Details
| Gene Symbol: Rmnd5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024288.2 | NP_077250.2 | 5 | 1085 | ||
| GenBank mRNA | AK003512.2 | - | 1 | 17 | ||
| AK148556.1 | - | 4 | 320 | |||
| AK149170.1 | - | 4 | 320 | |||
| AK164384.1 | - | 5 | 1086 | |||
| BC034085.1 | - | 1 | 71 | |||
| BC050876.1 | - | 5 | 936 | 6512227 | ||

Rmnd5a
Genomic Map