siRNA Details
| Gene Symbol: Utp11l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026031.3 | NP_080307.1 | 7 | 639 | ||
| GenBank mRNA | AK007361.1 | - | 5 | 410 | ||
| AK008801.1 | - | 3 | 191 | |||
| AK012550.1 | - | 7 | 640 | |||
| AK145266.1 | - | 7 | 672 | |||
| AK147113.1 | - | 7 | 670 | |||
| BC043139.1 | - | 5 | 3728 | |||
| BC061074.1 | - | 7 | 668 | 6706808 | ||

Utp11l
Genomic Map