siRNA Details
| Gene Symbol: Mboat2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001083341.1 | NP_001076810.1 | 2 | 224 | ||
| NM_026037.3 | NP_080313.2 | 2 | 224 | |||
| GenBank mRNA | AB297383.1 | - | 2 | 123 | ||
| AB305046.1 | - | 2 | 123 | |||
| AK031824.1 | - | 2 | 209 | |||
| AK076045.1 | - | 2 | 214 | |||
| AU067514.1 | - | 2 | 224 | |||
| BC025020.1 | - | 2 | 174 | 5364068 | ||

Mboat2
Genomic Map