siRNA Details
| Gene Symbol: ST6GALNAC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006456.2 | NP_006447.2 | 1, 2 | 388 | ||
| XM_005256954.3 | XP_005257011.1 | 1, 2 | 391 | |||
| GenBank mRNA | AJ251053.1 | - | 1, 2 | 172 | ||
| AK127567.1 | - | 1 | 2175 | |||
| AK225943.1 | - | 1, 2 | 186 | |||
| BC040455.1 | - | 1, 2 | 170 | 4252792 | ||
| BT019972.1 | - | 1, 2 | 133 | |||
| U14550.1 | - | 1, 2 | 172 | |||

ST6GALNAC2
Genomic Map