siRNA Details
| Gene Symbol: Dph7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026044.3 | NP_080320.1 | 5 | 542 | ||
| GenBank mRNA | AK013297.1 | - | 5 | 522 | ||
| AK078448.1 | - | 5 | 505 | |||
| AK087998.1 | - | 4 | 417 | |||
| AK152000.1 | - | 4 | 455 | |||
| AK153537.1 | - | 4 | 455 | |||
| BC056962.1 | - | 3 | 1192 | |||
| BC086486.1 | - | 5 | 544 | |||

Dph7
Genomic Map