siRNA Details
| Gene Symbol: Dph7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026044.3 | NP_080320.1 | 9 | 1745 | ||
| GenBank mRNA | AK013297.1 | - | 9 | 1724 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dph7| Gene Symbol: Dph7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026044.3 | NP_080320.1 | 9 | 1745 | ||
| GenBank mRNA | AK013297.1 | - | 9 | 1724 | ||