siRNA Details
| Gene Symbol: Zfp422 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF281634.1 | - | 2 | 931 | ||
| AK003498.1 | - | 1 | 103 | |||
| AK082970.1 | - | 2 | 835 | |||
| AK152681.1 | - | 2 | 819 | |||
| AK169251.1 | - | 2 | 1081 | |||
| BC018339.1 | - | 1 | 804 | 3583940 | ||

Zfp422
Genomic Map