siRNA Details
| Gene Symbol: Zfp422 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF281634.1 | - | 2 | 1366 | ||
| AK003498.1 | - | 1 | 537 | |||
| AK082970.1 | - | 2 | 1269 | |||
| AK152681.1 | - | 2 | 1253 | |||
| AK169251.1 | - | 2 | 1515 | |||
| BC018339.1 | - | 1 | 1238 | 3583940 | ||

Zfp422
Genomic Map