siRNA Details
| Gene Symbol: 2900005J15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK076604.1 | - | 1 | 1415 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | NM_026059.1 | - | 1415 | Chr. - - [-] on Build | ||

2900005J15Rik
Genomic Map