siRNA Details
| Gene Symbol: 3110040N11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026077.3 | NP_080353.1 | 5 | 765 | ||
| GenBank mRNA | AK014163.1 | - | 5 | 745 | ||
| AK019261.1 | - | 5 | 765 | |||
| AK170168.1 | - | 5 | 782 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

3110040N11Rik| Gene Symbol: 3110040N11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026077.3 | NP_080353.1 | 5 | 765 | ||
| GenBank mRNA | AK014163.1 | - | 5 | 745 | ||
| AK019261.1 | - | 5 | 765 | |||
| AK170168.1 | - | 5 | 782 | |||