siRNA Details
| Gene Symbol: Zc3h13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026083.2 | NP_080359.2 | 2 | 377 | ||
| GenBank mRNA | AK013285.1 | - | 2 | 379 | ||
| AK014200.1 | - | 2 | 157 | |||
| AK078225.1 | - | 2 | 368 | |||
| BB619434.1 | - | 2 | 378 | |||
| BC138264.1 | - | 2 | 326 | 8861283 | ||

Zc3h13
Genomic Map