siRNA Details
| Gene Symbol: Herc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026101.4 | NP_080377.2 | 4 | 841 | ||
| NM_030114.2 | NP_084390.1 | 4 | 841 | |||
| GenBank mRNA | AK009436.1 | - | 4 | 696 | ||
| AK019546.1 | - | 4 | 837 | |||
| AK143111.1 | - | 4 | 715 | |||
| AK147723.1 | - | 4 | 728 | |||
| BC043082.1 | - | 4 | 653 | 6417111 | ||
| BC060033.1 | - | 2 | 426 | 6478123 | ||

Herc4
Genomic Map