siRNA Details
| Gene Symbol: Bbs2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026116.2 | NP_080392.1 | 7 | 1193 | ||
| GenBank mRNA | AF342737.1 | - | 7 | 1193 | ||
| AK010779.1 | - | 7 | 806 | |||
| BC057184.1 | - | 7 | 852 | 5066846 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bbs2| Gene Symbol: Bbs2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026116.2 | NP_080392.1 | 7 | 1193 | ||
| GenBank mRNA | AF342737.1 | - | 7 | 1193 | ||
| AK010779.1 | - | 7 | 806 | |||
| BC057184.1 | - | 7 | 852 | 5066846 | ||