siRNA Details
| Gene Symbol: Fundc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026126.4 | NP_080402.3 | 4 | 384 | ||
| GenBank mRNA | AK013460.1 | - | 4 | 362 | ||
| AK014705.1 | - | 4 | 361 | |||
| AK031214.1 | - | 4 | 361 | |||
| AY234858.1 | - | 4 | 350 | |||
| BC042794.1 | - | 3 | 341 | |||
| BC049566.1 | - | 4 | 359 | 6742356 | ||
| BC061464.1 | - | 4 | 354 | 6819539 | ||

Fundc2
Genomic Map