siRNA Details
| Gene Symbol: 4833420G17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113550.1 | NP_001107022.1 | 10 | 2013 | ||
| NM_026127.4 | NP_080403.4 | 11 | 2076 | |||
| GenBank mRNA | AK146515.1 | - | 10 | 2004 | ||
| AK160863.1 | - | 10 | 2236 | |||
| BC021327.1 | - | 11 | 2056 | 5066171 | ||
| BC028328.1 | - | 11 | 2845 | |||

4833420G17Rik
Genomic Map