siRNA Details
| Gene Symbol: 4921536K21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026150.3 | NP_080426.1 | 3 | 1683 | ||
| GenBank mRNA | AK015005.1 | - | 3 | 1656 | ||
| AK049235.1 | - | 3 | 1655 | |||
| AK162162.1 | - | 4 | 509 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4921536K21Rik| Gene Symbol: 4921536K21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026150.3 | NP_080426.1 | 3 | 1683 | ||
| GenBank mRNA | AK015005.1 | - | 3 | 1656 | ||
| AK049235.1 | - | 3 | 1655 | |||
| AK162162.1 | - | 4 | 509 | |||