siRNA Details
| Gene Symbol: Pkp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026163.2 | NP_080439.1 | 13 | 2525 | ||
| GenBank mRNA | AK004650.1 | - | 13 | 2526 | ||
| AK049481.1 | - | 13 | 2526 | |||
| BC018487.1 | - | 7 | 1082 | |||
| BC037133.1 | - | 8 | 1131 | |||
| BC138252.1 | - | 13 | 2526 | 8861271 | ||
| BC138253.1 | - | 13 | 2525 | 8861272 | ||

Pkp2
Genomic Map