siRNA Details
| Gene Symbol: Poc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026173.3 | NP_080449.1 | 6 | 713 | ||
| GenBank mRNA | AK004769.2 | - | 6 | 641 | ||
| AK012926.1 | - | 6 | 681 | |||
| AK089013.1 | - | 6 | 649 | |||
| AK098149.1 | - | 6 | 606 | |||
| AK150656.1 | - | 6 | 714 | |||
| BC027118.1 | - | 6 | 651 | 5359153 | ||

Poc5
Genomic Map