siRNA Details
| Gene Symbol: Gpalpp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026177.3 | NP_080453.2 | 8 | 2724 | ||
| GenBank mRNA | AK004707.1 | - | 8 | 2704 | ||
| AK173238.1 | - | 8 | 2669 | |||
| BC029731.1 | - | 8 | 2685 | 4946215 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gpalpp1| Gene Symbol: Gpalpp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026177.3 | NP_080453.2 | 8 | 2724 | ||
| GenBank mRNA | AK004707.1 | - | 8 | 2704 | ||
| AK173238.1 | - | 8 | 2669 | |||
| BC029731.1 | - | 8 | 2685 | 4946215 | ||