siRNA Details
| Gene Symbol: Gpatch1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026181.1 | NP_080457.1 | 4 | 360 | ||
| GenBank mRNA | AK004868.1 | - | 4 | 360 | ||
| AK154571.1 | - | 4 | 341 | |||
| BC100577.1 | - | 4 | 335 | |||
| BC125584.1 | - | 4 | 351 | 40130099 | ||
| BC132155.1 | - | 4 | 351 | 40130432 | ||

Gpatch1
Genomic Map