siRNA Details
| Gene Symbol: Slc47a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026183.5 | NP_080459.2 | 15, 16 | 1440 | ||
| GenBank mRNA | AK004994.2 | - | 16, 17 | 1796 | ||
| AK009038.1 | - | 10, 11 | 1060 | |||
| BC031436.1 | - | 15, 16 | 1424 | |||

Slc47a1
Genomic Map