siRNA Details
| Gene Symbol: Ero1lb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026184.2 | NP_080460.2 | 12 | 1034 | ||
| GenBank mRNA | AF489856.1 | - | 12 | 840 | ||
| AK004981.1 | - | 1011 | ||||
| AK041951.1 | - | 12 | 1010 | |||
| AK160664.1 | - | 12 | 1050 | |||
| BC058721.1 | - | 12 | 1034 | 5705507 | ||
| BC111900.1 | - | 12 | 1004 | 40062137 | ||
| BC112426.1 | - | 12 | 1004 | 40062136 | ||
| BC145888.1 | - | 12 | 985 | 40131191 | ||
| BC145890.1 | - | 12 | 985 | 40131193 | ||

Ero1lb
Genomic Map