siRNA Details
| Gene Symbol: Cwc25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026186.4 | NP_080462.2 | 10 | 1434 | ||
| GenBank mRNA | AK004983.2 | - | 10 | 1378 | ||
| AK017849.1 | - | 10 | 1374 | |||
| AK035158.1 | - | 10 | 1362 | |||
| BC053447.1 | - | 3 | 588 | |||
| BC079553.1 | - | 9 | 4106 | 6856335 | ||

Cwc25
Genomic Map