siRNA Details
| Gene Symbol: 1700028P14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026188.2 | NP_080464.1 | 2 | 300 | ||
| GenBank mRNA | AK006473.1 | - | 2 | 303 | ||
| AK014972.1 | - | 2 | 302 | |||
| BC049563.1 | - | 2 | 285 | 6771302 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700028P14Rik| Gene Symbol: 1700028P14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026188.2 | NP_080464.1 | 2 | 300 | ||
| GenBank mRNA | AK006473.1 | - | 2 | 303 | ||
| AK014972.1 | - | 2 | 302 | |||
| BC049563.1 | - | 2 | 285 | 6771302 | ||