siRNA Details
| Gene Symbol: Eepd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026189.3 | NP_080465.3 | 2 | 583 | ||
| GenBank mRNA | AA543616.1 | - | 2 | 327 | ||
| AK009180.2 | - | 2 | 578 | |||
| AK019086.1 | - | 2 | 578 | |||
| AK168564.1 | - | 2 | 584 | |||
| AK173239.1 | - | 2 | 633 | |||
| BC025571.1 | - | 2 | 647 | 5323841 | ||

Eepd1
Genomic Map