siRNA Details
| Gene Symbol: Eepd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026189.3 | NP_080465.3 | 6 | 1741 | ||
| GenBank mRNA | AK009180.2 | - | 6 | 1736 | ||
| AK019086.1 | - | 6 | 1741 | |||
| AK168564.1 | - | 6 | 1742 | |||
| AK173239.1 | - | 6 | 1791 | |||
| BC025571.1 | - | 6 | 1805 | 5323841 | ||
| CK619108.1 | - | 5 | 469 | |||

Eepd1
Genomic Map