siRNA Details
Gene Symbol: 1700019N19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026208.2 | NP_080484.1 | 6 | 913 | ||
GenBank mRNA | AK006135.1 | - | 6 | 914 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 1700019N19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026208.2 | NP_080484.1 | 6 | 913 | ||
GenBank mRNA | AK006135.1 | - | 6 | 914 |