siRNA Details
| Gene Symbol: Cog6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026225.3 | NP_080501.2 | 2 | 281 | ||
| GenBank mRNA | AB093286.1 | - | 2 | 253 | ||
| AK016663.1 | - | 2 | 294 | |||
| AK031149.1 | - | 2 | 275 | |||
| AK045223.1 | - | 2 | 275 | |||
| AK048986.1 | - | 2 | 268 | |||
| AK153320.1 | - | 2 | 270 | |||
| BC025427.1 | - | 2 | 287 | 2649589 | ||

Cog6
Genomic Map