siRNA Details
Gene Symbol: Gpr89 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026229.4 | NP_080505.1 | 5, 6 | 494 | ||
GenBank mRNA | AK002937.1 | - | 5, 6 | 514 | ||
AK009734.1 | - | 4, 5 | 373 | |||
AK016787.1 | - | 5, 6 | 494 | |||
AK034931.1 | - | 62 | ||||
BC010729.1 | - | 5, 6 | 487 | 3708143 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
